Imprimer
Tissue processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M525T
Fournisseur: Simport Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
M525-2T M525-3T M525-4T M525-5T M525-6T M525-7T M525-8T M525-9T M525-10T M525-11T M525-12T
SIMPM525-2TEA 219 EUR
SIMPM525-2T SIMPM525-3T SIMPM525-4T SIMPM525-5T SIMPM525-6T SIMPM525-7T SIMPM525-8T SIMPM525-9T SIMPM525-10T SIMPM525-11T SIMPM525-12T
Tissue processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M525T
Cassettes d’enrobage Cassettes de traitement pour tissus
These cassettes are moulded from acetal and keep specimens safely submerged in liquid, and are resistant to the chemical action of most histological solvents.
  • Anterior writing area at a 45° angle
  • Taped cassettes to be used with Leica ink jet printers
  • Easy to remove disposable cover
Unisette™ histology cassette comes with a 45° anterior writing surface. This configuration is considered ideal for optimal workflow in some automated cassette printers and efficient barcode reading.
The efficient flow-through slots maximise fluid exchange and ensure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. The snap-latch and hinge-lock design of the UNISETTE™ prevents early separation of base and lid and allows for one-handed operation.
Cdt: Cassettes are packed in stacks of 40. Each case contains 25 stacks for a total of 1000 cassettes.