Imprimer
Overalls, Tritex® standard white, with hood
Fournisseur: Zellstoff-Vertriebs-GmbH & Co. KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
30757L 30757XL 30757XXL 30757XXXL
ZVGA30757LEA 236 EUR
ZVGA30757L ZVGA30757XL ZVGA30757XXL ZVGA30757XXXL
Overalls, Tritex® standard white, with hood
Vêtements à usage unique
Designed to protect against light spray, liquid aerosol or low pressure, low volume splashes, and airborne solid particulates. This one-piece coverall is manufactured from non-woven SMS.
  • Attached hood
  • Zip front opening with self-adhesive flap
  • Elasticated cuffs, ankles, hood, and waist
  • Serged seams
Coveralls are disposable, and should not be washed, dryed, ironed, bleached, or dry cleaned.
Colour: White
Certifications: CE 0624, Cat. III, Type 5/6