Imprimer
Avantor® Hichrom HI-DEX B-SE, GC Columns
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HI72-25-015-15EA 1250 EUR
HI72-25-015-15 HI72-25-015-25 HI72-25-015-30 HI72-25-015-50 HI72-25-015-60 HI72-25-025-15 HI72-25-025-25 HI72-25-025-30 HI72-25-025-50 HI72-25-025-60 HI77-10-010-10 HI77-10-010-5
Avantor® Hichrom HI-DEX B-SE, GC Columns
Colonnes pour chromatographie
Unique selectivity for the separation of enantiomers. Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.