Imprimer
Avantor® Hichrom HI-DEX DET Beta, GC Columns
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HI68-25-015-15EA 873 EUR
HI68-25-015-15 HI68-25-015-25 HI68-25-015-30 HI68-25-015-50 HI68-25-015-60 HI68-25-025-15 HI68-25-025-25 HI68-25-025-30 HI68-25-025-50 HI68-25-025-60 HI80-10-010-10 HI80-10-010-5
Avantor® Hichrom HI-DEX DET Beta, GC Columns
Colonnes pour chromatographie
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.