Imprimer
Labels for BBP33 printers, type B-724
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
133872 140723 140724 152847 152842 152913 152909 152910 152911 152908 152912 152840
BRDY133872EA 446 EUR
BRDY133872 BRDY140723 BRDY140724 BRDY152847 BRDY152842 BRDY152913 BRDY152909 BRDY152910 BRDY152911 BRDY152908 BRDY152912 BRDY152840
Labels for BBP33 printers, type B-724
Étiquettes
Amber high temperature polyimide with an ultra-durable adhesive withstands the extreme wash protocol and cleaning chemicals used in printed circuit board manufacturing.
  • Resists high temperatures up to 300 °C
  • Survives on the top or bottom side of printed circuit boards through the solder process
  • Service temperature: –70 to 270 °C
Withstands multiple cycles of harsh condition washes for printed circuit boards and electronic component preprocess labelling.
Certifications: Fulfills requirements for MIL-STD-202G method 215K and SAE AS81531 marking of electrical insulating material when used with R4300 series ribbons.