Imprimer
Spill kit, spill box
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
813910 813909 813908
BRDY813910EA 61.6 EUR
BRDY813910 BRDY813909 BRDY813908
Spill kit, spill box
Matériaux absorbants
These Spill box pads in a convenient dispenser box allows to create multiple spill stations.
  • Reusable plastic box can be refilled once all the sorbents have been used
  • The bright yellow colour stands out for quick identification and use
The Spill Box is light enough to be hanged to laboratory and storage area walls or to be attached to a forklift. Various application areas: in liquid storage areas, behind the seat of a truck, lab environments, in workstations that deal with liquids.