Imprimer
Pipe marker, water
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
N003371 N003412 N003349 N005832 N005873 N006135 N006160 N006279
BRDYN003371EA 16.8 EUR
BRDYN003371 BRDYN003412 BRDYN003349 BRDYN005832 BRDYN005873 BRDYN006135 BRDYN006160 BRDYN006279
Pipe marker, water
Panneaux
Individual markers on card with precut arrows, without pictogram.
  • Colour: White on green
  • Permanent acrylic
  • Pipe diameter: 20 to 49 mm