Imprimer
Workstation product and wire identification software suite
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
149191
BRDY149191EA 292 EUR
BRDY149191
Workstation product and wire identification software suite
Logiciels Logiciels de laboratoire
When you need to create product-identification and wire-marking labels, the product and wire identification suite is helpful to give solution.
  • Hardware: 1 gigahertz (GHZ) processor or faster
  • RAM: 2GB
  • Screen resolution: 1024×768 or higher (recommended), 800×600 high colour (minimum)
It allows you to create basic text labels or labels with graphics and custom formatting, and serialise and import data.
Operating system: Windows® 10;Windows® 7 (32- or 64-bit);Windows® 7 Enterprise;Windows® 7 Professional;Windows® 7 SPI;Windows® 7 Ultimate;Windows® 8 (32- or 64-bit);Windows® 8 Professional (32 or 64 bit);Windows® 8.1 (32- or 64-bit);Windows® 8.1 Enterprise;Windows® 8.1 Professional.
Language available: Bulgarian, Chinese (Simplified), Chinese (Traditional), Croatian, Czech, Danish, Dutch, English (US), Estonian, Finnish, French (France), German, Hungarian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese (Portugal), Romanian, Russian, Slovak, Slovenian, Spanish (Mexico), Spanish (Spain), Swedish, Turkish, Vietnamese.