Imprimer
Labels for BBP33 printers, type B-430
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
142886 142885 133887
BRDY142886EA 209 EUR
BRDY142886 BRDY142885 BRDY133887
Labels for BBP33 printers, type B-430
Labels
Clear polyester (B-430) labels are designed for applications that use barcodes, alphanumerics, graphic symbols and logos and require nameplate-like quality.
  • Excellent resistance to most chemicals, oils and water
  • Service temperature of –56 to +137 °C
Glossy, clear labels are thermal transfer printable and feature a durable permanent acrylic adhesive.
Certifications: UL approved; CSA accepted.