Imprimer
Label printer, BSP61
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
710949 711040
BRDY710949EA 4310 EUR
BRDY710949 BRDY711040
Label printer, BSP61
Printers Label Printers
The BSP™61 Print and apply system offers a fully automatic labelling solution anywhere along the production line. It combines a thermal transfer label printer with an automatic applicator and is designed to consistently print and accurately position and apply labels in almost all applications.
  • Print speed is 250 mm/s
  • Internal memory capacity is 64 mb with RAM/flash of 8 mb
  • Monocolour
  • Designed to handle up to 7000 labels per day
It remove the problems of automating label application to ensure reliable tracking and tracing with no cutting into the liner, clean pick up, no adhesive bleeding, no label curling and accuracy.
Higher volume applications such as PCB marking, component and product marking in high quality demanding industries like automotive, electronics and manufacturing.
Label dimension limits: Maximum width of 114 mm, minimum width of 10 mm, minimum height of 8 mm.
Informations de commande: For more information on available applicators or other accessories; contact your Avantor representative.