Imprimer
Thermal transfer ribbon for printer, R6400 series
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
066263 066264 620365
BRDY066263EA 140 EUR
BRDY066263 BRDY066264 BRDY620365
Thermal transfer ribbon for printer, R6400 series
Rubans pour imprimante
Ultra high chemical resistance at high temperatures. Superior solvent resistance.
  • Core - diameter (mm): 25.40 mm
  • Ribbon formulation: resin