Imprimer
Pots with lid, stainless steel
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A284-1250 A284-2500 A284-400 A284-5000 A284-650
300-0654EA 102 EUR
300-0654 300-0655 300-0656 300-0657 300-0658
Pots with lid, stainless steel
Plateaux/Récipients/Pots Pots
These stainless steel pots with lids are general purpose containers that are ideal for use with powders and liquids. The lid is a simple drop on design.
  • 304 stainless steel
  • Hygienic easy clean construction
  • Robust design
  • Easy to use
Ideal for use in food, chemical and general industry.