Imprimer
GMP PP tubs, white, SteriWare®
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8185P-250 8185P-500
300-0546EA 300 EUR
300-0546 300-0547
GMP PP tubs, white, SteriWare®
Récipients de collecte d'échantillon Conteneurs à usage universel
The polypropylene screw top containers are ideal for multiple applications including production, warehouse, sampling, and general laboratory work.
  • Robust polypropylene construction
  • Both EU and FDA compliant
  • Available in two volumes; 250 ml and 500 ml
  • Individually bagged and produced in a cleanroom
Tubs are manufactured in a cleanroom to ensure cleanliness and purity. The wide neck design makes them ideal for use with powders and creams.
Certifications: Containers conform to the latest standards including FDA, EU 10/2011, EC 1935/2004, and are BSE/TSE free.