Imprimer
Industrial gloves, HyFlex® 11-300
Fournisseur: Ansell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
11-300/9 11-300/7 11-300/8 11-300/11 11-300/10
ANSE11-300/9EA 28.8 EUR
ANSE11-300/9 ANSE11-300/7 ANSE11-300/8 ANSE11-300/11 ANSE11-300/10
Industrial gloves, HyFlex® 11-300
Gants
Glove liners made of high-elasticity polyamide for optimal fit.
  • Glove knitted with a seamless non-stop process
  • Excellent comfort and dexterity
  • Ambidextrous to increase wear life
  • Low-lint glove
Recommended for light duty tasks and food applications.
Certifications: Food contact approved
Cdt: 12 pairs per bag.