Imprimer
Tissue-clearing reagent, CUBIC-X1 for animals
Marques : TCI
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
T3866-25ML T3866-100ML
TCIAT3866-25MLEA 68.3 EUR
TCIAT3866-25ML TCIAT3866-100ML
Tissue-clearing reagent, CUBIC-X1 for animals
Colorants Réactifs pour la microscopie et la coloration
Specifications
Appearance: Colorless to Almost colorless clear liquid
Functionality test: to pass test (Remaining fluorescence strength)