Imprimer
α-Toluenesulfonyle fluorure, cristaux aiguilles blancs à blanc cassé
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Danger
Synonyme(s): PMSF, Fluorure de toluène-α-sulfonyle
02195381.2 0219538180 0219538101 0219538105 0219538125
ICNA02195381.2EA 25 EUR
ICNA02195381.2 ICNA0219538180 ICNA0219538101 ICNA0219538105 ICNA0219538125
α-Toluenesulfonyle fluorure, cristaux aiguilles blancs à blanc cassé
α-Toluenesulfonyle fluorure
Phenylmethylsulfonyl fluoride is an irreversible inhibitor of serine proteases - inhibits trypsin, chymotrypsin and protease. Inhibits mammalian acetyl cholinesterase, but not those from fish sources.
  • Presentation: White to off white needle crystals
  • Causes sulfonylation of the active-site serine residue
  • A less toxic substitute for the extremely toxic diisopropylfluorophosphate
Phenylmethylsulfonyl fluoride inhibits serine proteases such as trypsin and chymotrypsin. Also inhibits cysteine proteases (reversible by reduced thiols) and mammalian acetylcholinesterase.
Administration of PMSF produces analgesia unrelated to its anticholinesterase effect, and prolongs the analagesic effect of centrally administered β-endorphin.
Soluble in methanol and ethanol
Formule: C₇H₇FO₂S
Point d'ébullition: 80 - 85 °C (0,3 -rr)
Point de fusion: 89…99 °C
Température de stockage: Réfrigérateur
Numéro MDL: MFCD00007424
Numéro CAS: 329-98-6
EINECS: 206-350-2
UN: 2928
ADR: 6.1,II
Commander maintenant
Résultats des tests de spécification
Identity Test Passes
FTIR Conforms to Standard