Imprimer
Liquid samplers, Liquid Master
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5035T-1001 5035T-1501 5035T-2001 5035T-2501 5035T-601
300-0467EA 855 EUR
300-0467 300-0468 300-0469 300-0470 300-0471
Liquid samplers, Liquid Master
Échantillonneurs Préleveurs de liquides
An ideal tool for taking point samples from containers, drums, barrels, lakes, and tanks.
  • Easy clean design
  • High chemical resistance
  • Suitable for containers with an opening larger than 34 mm
  • Made from high quality USP class VI, FDA and EC conforming materials
The Liquid Master is manufactured from PTFE and FEP, which gives it excellent chemical resistance. Simple one-handed operation allows users to take liquid samples of a wide variety of liquids and chemicals.
The high specification USP Class VI material means it is ideal for pharma, food, and general sampling.