Imprimer
VWR® ARISTAR®, Cobalt Standard Solution, 10000 mg/L Co in dil. Nitric Acid (from Co), Standard for ICP
Marques : VWR Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
455274BEA 1480 EUR
455274B 455272W
VWR® ARISTAR®, Cobalt Standard Solution, 10000 mg/L Co in dil. Nitric Acid (from Co), Standard for ICP
Standards Cationic Standards Cobalt Standards
Co in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.