Imprimer
VWR®, Microseringues standard
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
549-0199EA 44.7 EUR
549-0199 549-0522 549-0523 549-0524
VWR®, Microseringues standard
Seringues Seringues de chromatographie
Standard micro syringes for GC with fixed needle.
  • All parts in contact with liquid are made of stainless steel and neutral borosilicate glass
Length in ordering table refers to needle length.