Imprimer
Lens cleaning tissues, grade 541
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
111-5003EA 26.1 EUR
111-5003 111-5004
Lens cleaning tissues, grade 541
Wipes Disposable Wipes and Towels Lens Papers
Suitable for cleaning various optical lenses such as objectives, binoculars, magnifying glasses, spectacles, as well as optical fibre filaments.
Thickness: 0,04 mm
Weight: 12 g/m²