Imprimer
VWR® Science Education Precision Balances
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
611-3236EA 583 EUR
611-3236 611-3260 611-3299 611-3468 611-3476 611-2602 611-2604 611-2607 611-3860
VWR® Science Education Precision Balances
Balances and Scales
Competitively priced electronic balances for the education sector. All models have a stainless steel top pan.
  • Easy to use
  • Small footprint
  • Selectable measuring units
  • Mains supply or optional external rechargeable battery for portable use
Additional benefits of the models (except 611-2605, 611-2606, 611-2607, 611-3236, 611-3246, 611-3299 and 611-3344) include:
RS232 serial interface
Hook for under-balance weighing suitable for external calibration models only (not for model number ending in i)
Capable of liquid and solid density determination, check-weighing, weight totalising, piece-counting, animal weighing, percentage weighing and peak hold functions
Balances, except those listed above, are compatible with the optional weighing tablet system
All these balances are now supplied without an internal battery to meet recent transport regulations. An optional external rechargeable battery (611-8684) may be purchased for portable operation.
User can choose three options for models with internal calibration (denoted by 'i' at end of the model name):
Balance will calibrate using the internal weight by pressing CAL button
Balance will automatically recalibrate when ambient temperature changes and/or after a certain time has passed (Incal+)
The user can choose to calibrate the balance with an external weight
Certifications: EN 61326-1:2013, EN 61010-1: 2010, EN 61010-2-010:2014, EN 50581:2012
Informations sur les accessoires: Optional Tablet System (611-8683) is suitable for use with models denoted with *.
Informations de commande: These balances are also available with a calibration certificate. Please contact your local Avantor sales office for details of this option.
Informations de livraison: Supplied with plastic in use protective cover.
Attention: -C balances are supplied with a factory calibration certificate.
Models with suffix G have large graphic display.