Imprimer
Sets de tampons absorbants
Fournisseur: Cytiva (Formerly Pall Lab)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
66025 66190
516-9012EA 311 EUR
516-9012 516-7850
Sets de tampons absorbants
Lingettes Lingettes nettoyantes
Handy dispenser kits allow one-handed dispensing of cellulose absorbent pads.
  • One-handed operation
  • Holds up to 100 pads
  • Sterile pads are sterilized by gamma irradiation
  • No hazardous EtO residuals
Dispense a clean cellulose pad into a petri dish without touching the pad.
Dispenser holds one tube of 100 absorbent pads. Tubes drop quickly into the hand dispenser for easy use. Absorbent pads are ideal for absorbing 2 ml of broth media to culture colonies in accordance with the membrane filter test method.
Pad Ø: 45.5 mm (1³/₄").
Pad thickness: 35 mil.
Informations de commande: Each kit includes one dispenser and 10 tubes, 100 pads per tube.