Imprimer
PC platinum certified clean biotainer bottles, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CE-N3030-42 CE-N3120-42 CE-N3233-42 CE-N3405-42 CE-N3410-42 CE-N3500-05 CE-N3500-20
216-2316EA 2307 EUR
216-2316 216-2317 216-2318 216-2319 216-2320 216-2323 216-2325
PC platinum certified clean biotainer bottles, Nalgene®
Flacons Flacons en plastique
The Nalgene™ multi-tiered portfolio provides bioproduction facilities with a variety of high quality, sterile storage and transport solutions. Choose the appropriate product necessary to suit any application, through the step-down process and into the cleanroom.
  • Durable, translucent PC construction ensures the integrity of samples
  • Closures feature a silicone liner for leakproof assurance
  • Customisation options are available for fluid transfer applications
  • Extensive validation documentation support
  • Suitable for use in storage conditions of –100 to 100 °C
Bottles are washed with USP water for injection and handled post-washing in Class 5 cleanrooms with Class 4 packaging/inspection hoods to produce these very low particulate products. Certified and lot-to-lot tested to contain less than ¹/₃ the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready to use products with a Sterility Assurance Level of 10⁻⁶ inside the container/closure, including two heat-sealed sterile barriers to protect the product prior to use.
Certifications: USP Class VI; USP 661; FDA 21CFR 177.1315.