Imprimer
Tyramide amplification kit with HRP goat, anti-mouse and CF® dye or biotin tyramide
Marques : Biotium
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
33000 33003 33006 33009 33012 33015 33018
33000.EA 842 EUR
33000. 33003. 33006. 33009. 33012. 33015. 33018.
Tyramide amplification kit with HRP goat, anti-mouse and CF® dye or biotin tyramide
Systèmes de liaison et de marquage des protéines
Kits for increasing immunofluorescence sensitivity using HRP-catalysed tyramide signal amplification with the choice of next-generation fluorescent CF® dyes or biotin.
  • Increase sensitivity by up to 100-fold
  • Use lower primary antibody concentrations
  • Boost signal in challenging samples, like human FFPE tissues
  • Choose from six bright and photostable CF® dyes or biotin
Tyramide amplification, sometimes called Catalysed Reporter Deposition (CARD), is an enzyme-mediated detection method that uses the catalytic activity of horseradish peroxidase (HRP) to generate high density labelling of a target protein or nucleic acid sequence in situ. The tyramide amplification method has been reported to increase the sensitivity by up to 100-fold compared to conventional avidin-biotin detection. Multiple rounds of tyramide signal amplification can be performed for multiplex detection.
The HRP goat anti-mouse antibody provided in the kits has been adsorbed against bovine, horse, human, rabbit, and swine serum to minimise cross-reactivity; the antibody is not cross-adsorbed against rat. The kits include your choice of six CF® dye tyramides or biotin tyramide. CF® dyes are the line of next-generation fluorescent dyes, with advantages in brightness and photostability compared to other dyes.
Informations de commande: Each kit contains reagents for staining 50 to 150 slides; the actual number of samples that can be stained will depend on staining volumes used. Tyramide amplification buffer, CF® Dye Tyramides, and other tyramide compounds are also available separately. Note: Endogenous biotin blocking reagents (not provided) may be required for streptavidin/biotin detection in cells or tissues.
Informations de livraison: Kit components: CF® dye or biotin-tyramide stock solution, HRP Goat anti-mouse IgG (highly cross-adsorbed), BSA for blocking, Tyramide amplification buffer plus, 30% Hydrogen peroxide.