Imprimer
DEC-QUAT® 100 Disinfectant
Fournisseur: VELTEK ASSOCIATES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DQ100-03-8Z-E DQ100-03-2Z-E DQ100-05-2Z-E DQ100-01-E DQ100-02-E DQ100-04-2Z-E
VELTDQ100-03-8Z-EEA 687 EUR
VELTDQ100-03-8Z-E VELTDQ100-03-2Z-E VELTDQ100-05-2Z-E VELTDQ100-01-E VELTDQ100-02-E VELTDQ100-04-2Z-E
DEC-QUAT® 100 Disinfectant
Produits nettoyants Désinfectants
DEC-QUAT® 100 is a quaternary ammonium solution for use on hard non-porous environmental surfaces.
  • Effective as a broad spectrum hospital disinfectant, fungicide, deodorizer, virucide
  • Food and non-food contact sanitizer effective against a broad spectrum of pathogenic bacteria
  • Available as both sterile and non-sterile
  • Completely traceable and has been validated for sterility and shelf life
This product is designed for use in hospital, medical and health care institutions as a broad-spectrum disinfectant solution on hard, non-porous environmental surfaces such as walls, ceilings, floors, and countertops. Effective against Pseudomonas aeruginosa, Staphylococcus aureus, and Salmonella choleraesuis. Effective against Hepatitis B Virus when contact time is 10 minutes at 1 oz:1 gal of water. Effective against Human Immunodeficiency Virus Type 1 (HIV) after 30 seconds of contact time.
Cdt: Filled sterile by filtration at 0.2 microns. Gamma-irradiated, double-bagged and delivered with lot-specific sterile documentation.