Imprimer
ESD safety shoes, lace up, Ambition Low
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
72733/36 72733/37 72733/38 72733/39 72733/40 72733/41 72733/42 72733/43 72733/44 72733/45 72733/46 72733/47 72733/48
ELTN72733/36EA 85.9 EUR
ELTN72733/36 ELTN72733/37 ELTN72733/38 ELTN72733/39 ELTN72733/40 ELTN72733/41 ELTN72733/42 ELTN72733/43 ELTN72733/44 ELTN72733/45 ELTN72733/46 ELTN72733/47 ELTN72733/48
ESD safety shoes, lace up, Ambition Low
Chaussures et bottes
Sporty and lightweight low-cut ESD safety shoes offer excellent wearing comfort, with a padded upper edge to protect the Achilles tendon and a full, padded bellows tongue to prevent pressure marks and dirt incursion.
  • Nubuck leather/mesh material
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Semi-orthopaedic inlay sole ESD grey
  • Soft fleece ESD insole
  • TPU/PU sole DIMENSION PRO
  • Steel toe cap
The Nubuck leather/mesh material construction results in a wear-resistant, tear-resistant, shape-retaining, quick drying, lightweight shoe that will provide comfort over extended periods of use. Ideally suited for use in dry work areas. Thanks to the excellent discharge capabilities, they are suitable for work in ESD sensitive or electrostatically protected areas (EPA).
Certifications: Comply to the standard 61340-5-1; EN ISO 20345 S1 SRC, form A