Imprimer
DEC-SPORE 200 plus, disinfectant and sporicide
Fournisseur: VELTEK ASSOCIATES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DS2000616Z01E DS200-04A-E DS200-05A-E DS200-01A-E-8 DS2000716Z01E DS2000313ZAE
VELTDS2000616Z01EEA 884 EUR
VELTDS2000616Z01E VELTDS200-04A-E VELTDS200-05A-E VELTDS200-01A-E-8 VELTDS2000716Z01E VELTDS2000313ZAE
DEC-SPORE 200 plus, disinfectant and sporicide
Produits nettoyants Désinfectants
DEC-SPORE 200 Plus is a sterilant-virucide-disinfectant-sanitizer. A peracetic acid hydrogen peroxide solution that is processed to comply with the standards required by pharmaceutical, biotechnology, medical device, and health care industries.
  • Filled in ISO 5 (Grade A/B, Former Class 100)
  • Aseptically filled and packaged into sterile components
  • Filtered at 0.2 microns
  • Sterility tested according to current USP compendium
  • Completely traceable from start to finish
  • Completely validated for sterility and shelf life
DEC-SPORE 200 Plus can be mixed to two different concentrations, disinfectant and sporicide. DEC-SPORE 200 Plus is available in multiple container sizes and in our patented SimpleMix system that allows for safe and fresh dilutions every single time.
Informations de commande: Available in sterile or non sterile options.