Imprimer
Ruban de masquage économique 101E
Fournisseur: 3M
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
101E-48MM 101E-18MM 101E-24MM 101E-30MM
MMMA101E-48MMEA 7.4 EUR
MMMA101E-48MM MMMA101E-18MM MMMA101E-24MM MMMA101E-30MM
Ruban de masquage économique 101E
Rubans adhésifs
This is a smooth, multi-purpose crepe paper tape for room temperature, indoor, light-duty applications. It is ideal for non-critical paint masking, but also bundling, sealing, attaching, and other applications.
  • Good initial tack to a wide variety of surfaces in different applications
  • Designed for light-duty applications
  • Operating temperature range up to 30 °C
  • Easy to write on for labelling
The cost-effective general-purpose value masking tape is a chamois-colored, 0,12 mm thick adhesive tape with rubber adhesive for general applications (temporary attachment, masking for painting without burning, etc.). It provides conformability on irregular surfaces and around corners, and is hand tearable for easy use.