Imprimer

Dextrose monohydrate, Conforme aux spécifications analytiques Ph. Eur., BP, Ph Franç. (7.0-9.5% H₂O), Fluka™

Marques : Honeywell Chemicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

16301H-6X2.5KG 16301H-1KG 16301H-2.5KG 16301H-6X1KG
HONC16301H-6X2.5KGEA 579 EUR
HONC16301H-6X2.5KG HONC16301H-1KG HONC16301H-2.5KG HONC16301H-6X1KG
Dextrose monohydrate, Conforme aux spécifications analytiques Ph. Eur., BP, Ph Franç. (7.0-9.5% H₂O), Fluka™
Dextrose monohydrate

  • Wide selection of support materials and multiple cartridge/column sizes
  • Every stationary phase batch tested to ensure lot-to-lot reproducibility
  • B&J's reliable quality

Point de fusion: 83 °C
Température de stockage: Température ambiante
Numéro CAS: 5996-10-1

Commander maintenant

Résultats des tests de spécification

Assay (HPLC, anhydrous substance) 97.5 - 102.0 %
Specific rotation C=10iNH₂O,calculatedto ANHydr.Substances 52.5 - 53.3 °
Conductivity 25 °C Max. 20 µS/cm
Water (Karl Fischer) 7.5 - 9.5 %
Sulfated ash Max. 0.1 %
Acid. or alk. react. impurities complying
Aluminium (Al) Max. 0.00005 %
Arsenic (As) Max. 1 ppm
Barium (Ba) complying
Calcium (Ca) Max. 200 ppm
Lead (Pb) Max. 0.5 ppm
Chloride (Cl) Max. 100 ppm
Sulfate (SO₄) Max. 200 ppm
Sulfite (as SO₂) Max. 10 ppm
Soluble starch, sulfite complying
Dextrines complying
Foreign sugars, soluble starch, dextrines complying
Related substances (HPLC) complying
Residual solvents complying
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR