Imprimer
Cleaning cloths, J-Cloth® Plus
Fournisseur: PGI NONWOVEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
74441 74442 74443 74444
115-2486EA 14.9 EUR
115-2486 115-2487 115-2488 115-2489
Cleaning cloths, J-Cloth® Plus
Wipes Reusable Cleaning Cloths Cloths for General Applications
Compostable J-Cloth® Plus cleaning wipes feature a unique, open, wavy texture that enables efficient pick-up of all types of dirt particles.
  • An effective green cleaning solution
  • Ensures environmentally aware waste management
  • Reduce waste and odours
  • Wipes are externally certified for FCC (food contact)
The unique open structure of each wipe allows for simple and thorough rinsing, ensuring wipes stay fresh for longer.
Certifications: Compostable in accordance with the DIN EN 13432 200-12 standard.