Imprimer
Stretchable silicone lids for laboratory vessels, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
291101119 291101127 291101135 291102115 291102123 291102131 291103111 291103128 291103136 291100011 291100028 291100036 291100044 291100052 291100069 291104116 291104124 291104132 291105112 291105129 291105137
213-0516EA 3.5 EUR
213-0516 213-0517 213-0518 213-0519 213-0520 213-0521 213-0522 213-0523 213-0524 213-0513 213-0514 213-0515 213-0525 213-0526 213-0527 213-0528 213-0529 213-0530 213-0531 213-0532 213-0533
Stretchable silicone lids for laboratory vessels, DURAN®
Capuchons Bouchons pour flacons
These stretchable silicone lids are used to close round and square vessels and are available in three sizes and colours, as well as a set of three.
  • For covering and sealing laboratory vessels
  • Lids are made of stretchable, chemical and heat resistant silicone
  • Recommended usable temperature range is –40 to +180 °C
  • Due to excellent stretchability, DURAN® silicone lids can be used for a wide variety of containers of different diameters and shapes
  • Lids may be sterilised (steam/autoclave), and are lab washer compatible
  • Each lid features a marking area
  • Reusable and easy to clean
The closure or covering of vessels is an everyday topic in the laboratory. Often glass plates or aluminium foil are used. From the point of view of environmental friendliness and durability, both variants may be classified as disadvantageous. The newly developed silicone cover is a good alternative to conventional cover variants and is available individually in three sizes or as a set. Due to its elasticity, the lid can be used for a variety of vessel openings, square or round. The silicone lid has a slightly larger tab that can be used as a writing field. The three colours offered are cyan, pink and green, and allow for convenient colour identification.
Max. operating temperature: 140 °C
Cdt: Individual lids are supplied one piece supplied per pack. Lid sets are supplied one piece of each size (S, M, L) per pack.