Imprimer
General purpose gloves, Showa 4400Y
Fournisseur: Showa
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4400Y-10/XL 4400Y-7/S 4400Y-8/M 4400Y-9/L
112-5830EA 82.4 EUR
112-5830 112-5831 112-5832 112-5833
General purpose gloves, Showa 4400Y
Gants
These flexible gloves with ¾ nitrile coating protect the hand from oils and abrasion without compromising sensitivity and flexibility.
  • Soft cotton jersey liner
  • Absorbs perspiration to increase comfort
  • Good mechanical resistance
  • Prevents oil and dirt penetration
  • Elasticated wrist cuff
For increased visibility and ease of identification, the 4400Y is coated in a fluorescent yellow nitrile.
Suitable for applications in agriculture, automotive, gardening, mechanical, warehousing and distribution.
Certifications: EN 388:2016 (3111X); Cat. II
Cdt: 12 pairs per polybag.