Imprimer
Liquid and cold resistant gloves, Showa 477
Fournisseur: Showa
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
477-7/M 477-8/L 477-9/XL 477-10/XXL
112-5850EA 1840 EUR
112-5850 112-5851 112-5852 112-5849
Liquid and cold resistant gloves, Showa 477
Gants
Designed to keep hands warm and dry in the most extreme environments, the 477 cold resistant glove combines thermal insulation with tactical grip. Foam grip and a dual nitrile coating ensure precision in wet and oily environments, whilst the acrylic liner provides protection from cold weather.
  • Dual nitrile coating technology provides long lasting grip
  • Seamless knit for optimum comfort
  • Well protected wrist
  • Innovative insulated double nitrile foam grip
  • Fixed acrylic terry liner
  • Impermeable
Ideal for agriculture, aerospace, construction, cold storage, forestry, fishing, railways, oil, gas, municipal services warehousing and distribution. Typical applications include cold storage, concreting, handling oily components in inclement conditions, refrigerated vehicles, ship maintenance.
Colour: White/blue/black
Certifications: EN 388:2016 (4231X); EN 511:2006 (011); Cat. II
Cdt: 6 pairs per bag, 12 bags per case.