Imprimer
Impact resistant gloves, showa 377-IP
Fournisseur: Showa
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
377IP-10/XXL 377IP-7/M 377IP-8/L 377IP-9/XL
112-5813EA 212 EUR
112-5813 112-5814 112-5815 112-5816
Impact resistant gloves, showa 377-IP
Gants
These nitrile foam Hi-Vis anti-impact gloves have a highly protective design to prevent injuries caused by impact and other hazards. The SHOWA-configured Thermo Plastic Rubber (TPR) shields to the back of the fingers and hand are designed to bolster protection against crushes and impacts.
  • Designed for easy movement and continuous wear
  • No irritation by seams in contact with the skin
  • Well protected wrist
  • Anatomical shape
  • Reinforced knit cuff
The SHOWA patented nitrile foam coating provides excellent grip in wet and greasy conditions, flat nitrile layers help protect against contaminants, fexible and durable TPR shields.
Typical applications include: Aerospace, agriculture, automotive, commercial fishing, construction, manufacturing, ports, railways, oil, gas warehousing and distribution.
Colour: White/blue/black/green
Certifications: EN 388:2016 (4121X); Cat. II
Cdt: 5 pairs per pack.