Imprimer
General purpose gloves, Showa 376R
Fournisseur: Showa
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
376R-9/XL 376R-6/S 376R-7/M 376R-8/L 376R-10/XXL
112-5807EA 112 EUR
112-5807 112-5804 112-5805 112-5806 112-5803
General purpose gloves, Showa 376R
Gants
These liquid-resistant gloves protect from liquid penetration with a double nitrile outer layer. The foam coating over nitrile ensures safety with an absorbent grip, even when working with oily components.
  • 3/4 nitrile dipped with extra foam nitrile on palm
  • Breathable and water resistant
  • High dexterity and resistance to wear and tear
  • Elasticated wrist cuff, scalloped edge
  • Anatomical shape with seamless knit
  • Minimal allergy risks
Protect the hand from oils, hydrocarbons, grease and abrasion, with optimal long lasting grip. Impermeable for working in damp or greasy environments, with increased resistance to abrasion for better durability.
Suitable applications: Aerospace, automotive, construction, manufacturing, oil and gas.
Colour: Dark grey/grey/blue/black.
Certifications: EN 388:2016 (4121X); Cat. II
Cdt: 10 pairs per pack.
Attention: Do not use in cases of heavy chemical exposure, electrical or thermal risk. Store away from light and humidity.