Imprimer
General purpose gloves, Showa 370
Fournisseur: Showa
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
370BLA-L 370BLA-M 370BLA-S 370BLA-XL 370BLA-XXL
112-5792EA 86.8 EUR
112-5792 112-5793 112-5794 112-5795 112-5796
General purpose gloves, Showa 370
Gants
This lightweight palm-coated nitrile glove is ideal for protecting hands against fluids such as oils. It allows finger dexterity to be maintained whilst being robust and hard-wearing.
  • Seamless nylon knit
  • Protects the hand from oil, hydrocarbon and grease penetration
  • 13 gauge seamless lint-free liner
  • Resistant to deformation
  • High levels of dexterity and tactility
  • Black colouring helps extend wearable life
Applications include assembling components, handling greasy components, inspection, machining, maintenance and plastic moulding.
Certifications: EN 388:2016 (4121X); Cat. II
Cdt: 10 pairs per pack.
Attention: Do not use in cases of chemical, electrical or thermal risk. Store away from light and humidity