Imprimer
VWR®, Synthetic Sea Water Analytical Reagent manufactured in accordance with ASTM D665 / IP 135
Marques : VWR Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
99169.001EA 216 EUR
99169.001 99169.251 99169.260 99169.360
VWR®, Synthetic Sea Water Analytical Reagent manufactured in accordance with ASTM D665 / IP 135
Étalons Étalons pour l'analyse environnementale
A high quality analytical reagent manufactured and certified compliant for use in ASTM D 665 / IP 135 the Standard Test Method for Rust-Preventing Characteristics of Inhibited Mineral Oil in the Presence of Water.
  • A Synthetic Sea Water Analytical Reagent manufactured in accordance with ASTM D665 / IP 135
  • Provided with a certificate which includes an estimation of uncertainty of manufacture
  • Ensures full compliance with ASTM and IP test method requirements for reagent preparation
  • Conveniently packaged in single dose or multi dose packs
  • Cost effective and time saving
  • Certificate of Manufacture and SDS included
  • 12 month shelf life
A high quality analytical reagent manufactured and certified compliant for use in ASTM D665 / IP 135 the Standard Test Method for Rust-Preventing Characteristics of Inhibited Mineral Oil in the Presence of Water. It should be noted that ASTM D1401, Standard Test Method for Water Separability of Petroleum Oils and Synthetic Fluids, permits the use of synthetic seawater as defined by ASTM D665, for certain types certain oils or fuels used in marine applications. Reference should be made to any manufacturer specifications for specific usage.