Imprimer
Industrial gloves, LIMA, TRIUS®
Fournisseur: TRIUS SAFETY
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
LIMA/10 LIMA/11 LIMA/7 LIMA/8 LIMA/9
TRIULIMA/10EA 21.6 EUR
TRIULIMA/10 TRIULIMA/11 TRIULIMA/7 TRIULIMA/8 TRIULIMA/9
Industrial gloves, LIMA, TRIUS®
Gants
Universal light nitrile coated glove designed for routine handling in wet or greasy environments.
  • Ultra-light product
  • Robust NBR coating
  • High dexterity
  • Good when handling greasy parts
These coated protective gloves are made of white polyester, with grey nitrile palm coating.
Certifications: EN 388:2016 (4123X), EN 420:2003 + A1:2009 dexterity 5, REACh compliant.
Cdt: Bundle of 10 pairs with UIS
Attention: Not for use in case of heat.