Imprimer
Industrial gloves, COSMOS, TRIUS®
Fournisseur: TRIUS SAFETY
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
COSMOS/8 COSMOS/6 COSMOS/7 COSMOS/10 COSMOS/11 COSMOS/9
112-6022EA 85.2 EUR
112-6022 112-6020 112-6021 112-6018 112-6019 112-6023
Industrial gloves, COSMOS, TRIUS®
Gants
This multi-purpose, grain and split combination cow leather glove is useful for a large variety of work.
  • High quality leather
  • Extra grip
  • Good dexterity
  • Elastic on back
These multi-purpose gloves are designed for use in dry environments. The handling glove features a natural cowhide grain leather palm, split leather back, and elasticated wrist band on the back.
Certifications: EN 388: 2016 3132X. EN 420:2003 + A1:2009 dexterity 5. REACh compliant.
Cdt: Bundle of 10 pairs with UIS, carton of 100 pairs.