Imprimer
ESD safety shoes, Boa® Fit System, uvex 1 x-tended support, 6567
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6567.1/35 6567.1/36 6567.1/37 6567.1/38 6567.1/39 6567.1/40 6567.1/41 6567.1/42 6567.1/43 6567.1/44 6567.1/45 6567.1/46 6567.1/47 6567.1/48 6567.1/49 6567.1/50 6567.1/51 6567.1/52 6567.2/35 6567.2/36 6567.2/37 6567.2/38 6567.2/39 6567.2/40 6567.2/41 6567.2/42 6567.2/43 6567.2/44 6567.2/45 6567.2/46 6567.2/47 6567.2/48 6567.2/49 6567.2/50 6567.2/51 6567.2/52 6567.3/35 6567.3/36 6567.3/37 6567.3/38 6567.3/39 6567.3/40 6567.3/41 6567.3/42 6567.3/43 6567.3/44 6567.3/45 6567.3/46 6567.3/47 6567.3/48 6567.3/49 6567.3/50 6567.3/51 6567.3/52
UVEX6567.1/35EA 140 EUR
UVEX6567.1/35 UVEX6567.1/36 UVEX6567.1/37 UVEX6567.1/38 UVEX6567.1/39 UVEX6567.1/40 UVEX6567.1/41 UVEX6567.1/42 UVEX6567.1/43 UVEX6567.1/44 UVEX6567.1/45 UVEX6567.1/46 UVEX6567.1/47 UVEX6567.1/48 UVEX6567.1/49 UVEX6567.1/50 UVEX6567.1/51 UVEX6567.1/52 UVEX6567.2/35 UVEX6567.2/36 UVEX6567.2/37 UVEX6567.2/38 UVEX6567.2/39 UVEX6567.2/40 UVEX6567.2/41 UVEX6567.2/42 UVEX6567.2/43 UVEX6567.2/44 UVEX6567.2/45 UVEX6567.2/46 UVEX6567.2/47 UVEX6567.2/48 UVEX6567.2/49 UVEX6567.2/50 UVEX6567.2/51 UVEX6567.2/52 UVEX6567.3/35 UVEX6567.3/36 UVEX6567.3/37 UVEX6567.3/38 UVEX6567.3/39 UVEX6567.3/40 UVEX6567.3/41 UVEX6567.3/42 UVEX6567.3/43 UVEX6567.3/44 UVEX6567.3/45 UVEX6567.3/46 UVEX6567.3/47 UVEX6567.3/48 UVEX6567.3/49 UVEX6567.3/50 UVEX6567.3/51 UVEX6567.3/52
ESD safety shoes, Boa® Fit System, uvex 1 x-tended support, 6567
Chaussures et bottes
Sporty, multifunctional safety shoes are available in multiple widths.
  • Especially lightweight and flexible
  • Breathable, water-resistant, high-tech microvelour upper material
  • Foamed polyurethane scuffcap helps increase product life of upper
  • Meets the ESD specification with volume resistance less than 35 megaohms
  • Boa® Fit System for an optimised fit, allowing precise and individual adjustment
The 100% metal-free uvex xenova® toe cap offers compact design, anatomical shape, good lateral stability, and no thermal conductivity properties. The metal-free, penetration-resistant insole is in accordance with the latest standard, which does not impact shoe flexibility. Safety support shoes feature medial and lateral heel support for the foot through a specially designed side frame to protect against ankle injuries and impacts. Suitable for people allergic to chrome, as they are made from synthetic materials. All sole materials are free of silicones, plasticisers, and other paint wetting impairment substances.
Colour: Black/red
Certifications: EN 61340 (ESD); EN ISO 20345: 2011 S3 SRC