Imprimer
ESD safety shoes, Boa® fit system, uvex 2 VIBRAM®, 6531
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6531.1/35 6531.1/36 6531.1/37 6531.1/38 6531.1/39 6531.1/40 6531.1/41 6531.1/42 6531.1/43 6531.1/44 6531.1/45 6531.1/46 6531.1/47 6531.1/48 6531.1/49 6531.1/50 6531.1/51 6531.1/52 6531.2/35 6531.2/36 6531.2/37 6531.2/38 6531.2/39 6531.2/40 6531.2/41 6531.2/42 6531.2/43 6531.2/44 6531.2/45 6531.2/46 6531.2/47 6531.2/48 6531.2/49 6531.2/50 6531.2/51 6531.2/52 6531.3/35 6531.3/36 6531.3/37 6531.3/38 6531.3/39 6531.3/40 6531.3/41 6531.3/42 6531.3/43 6531.3/44 6531.3/45 6531.3/46 6531.3/47 6531.3/48 6531.3/49 6531.3/50 6531.3/51 6531.3/52
UVEX6531.1/35EA 174 EUR
UVEX6531.1/35 UVEX6531.1/36 UVEX6531.1/37 UVEX6531.1/38 UVEX6531.1/39 UVEX6531.1/40 UVEX6531.1/41 UVEX6531.1/42 UVEX6531.1/43 UVEX6531.1/44 UVEX6531.1/45 UVEX6531.1/46 UVEX6531.1/47 UVEX6531.1/48 UVEX6531.1/49 UVEX6531.1/50 UVEX6531.1/51 UVEX6531.1/52 UVEX6531.2/35 UVEX6531.2/36 UVEX6531.2/37 UVEX6531.2/38 UVEX6531.2/39 UVEX6531.2/40 UVEX6531.2/41 UVEX6531.2/42 UVEX6531.2/43 UVEX6531.2/44 UVEX6531.2/45 UVEX6531.2/46 UVEX6531.2/47 UVEX6531.2/48 UVEX6531.2/49 UVEX6531.2/50 UVEX6531.2/51 UVEX6531.2/52 UVEX6531.3/35 UVEX6531.3/36 UVEX6531.3/37 UVEX6531.3/38 UVEX6531.3/39 UVEX6531.3/40 UVEX6531.3/41 UVEX6531.3/42 UVEX6531.3/43 UVEX6531.3/44 UVEX6531.3/45 UVEX6531.3/46 UVEX6531.3/47 UVEX6531.3/48 UVEX6531.3/49 UVEX6531.3/50 UVEX6531.3/51 UVEX6531.3/52
ESD safety shoes, Boa® fit system, uvex 2 VIBRAM®, 6531
Chaussures et bottes
No challenge is too great for the innovative uvex 2 safety shoe series in protection class S3. They offer ergonomic shape and optimum climate properties to ensure top performance for daily working life. Boa® Fit System for precision fit to ensure long-lasting comfort.
  • Lightweight and flexible safety shoe
  • Anti-slip VIBRAM® rubber soles
  • Soft padding on dust tongue and collar
  • Foamed polyurethane scuffcap helps increase product life of upper
  • Virtually seam-free construction to eliminate pressure points
  • Meets ESD specifications with leakage resistance under 35 megaohms
  • HRO symbol for heat-resistant substructure for brief exposure up to +300 °C
  • HI marking for heat-insulating substructure (tested in accordance with standard at +150 °C)
The 100% metal-free uvex xenova® toe cap offers compact design, anatomical shape, good side and ankle twist stability, no thermal conductivity, and anti-magnetic properties. Metal-free, penetration-resistance insole is in accordance with the latest standard, which does not impact shoe flexibility. Certified with uvex medicare for individual, orthopaedic inlay soles and individual adjustments of the sole unit such as overall increase, inside and outside increase, and other solutions. Removable anti-static insole, with moisture control system, provides additional shock absorption in the heel and at the front of the foot.
Colour: Black/orange
Certifications: EN 61340 (ESD); EN ISO 20345:2011, S3, SRC, HI, HRO