Imprimer
ESD safety shoes, Boa® Fit System, uvex 1 x-tended support, 6568
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6568.1/35 6568.1/36 6568.1/37 6568.1/38 6568.1/39 6568.1/40 6568.1/41 6568.1/42 6568.1/43 6568.1/44 6568.1/45 6568.1/46 6568.1/47 6568.1/48 6568.1/49 6568.1/50 6568.1/51 6568.1/52 6568.2/35 6568.2/36 6568.2/37 6568.2/38 6568.2/39 6568.2/40 6568.2/41 6568.2/42 6568.2/43 6568.2/44 6568.2/45 6568.2/46 6568.2/47 6568.2/48 6568.2/49 6568.2/50 6568.2/51 6568.2/52 6568.3/35 6568.3/36 6568.3/37 6568.3/38 6568.3/39 6568.3/40 6568.3/41 6568.3/42 6568.3/43 6568.3/44 6568.3/45 6568.3/46 6568.3/47 6568.3/48 6568.3/49 6568.3/50 6568.3/51 6568.3/52
UVEX6568.1/35EA 154 EUR
UVEX6568.1/35 UVEX6568.1/36 UVEX6568.1/37 UVEX6568.1/38 UVEX6568.1/39 UVEX6568.1/40 UVEX6568.1/41 UVEX6568.1/42 UVEX6568.1/43 UVEX6568.1/44 UVEX6568.1/45 UVEX6568.1/46 UVEX6568.1/47 UVEX6568.1/48 UVEX6568.1/49 UVEX6568.1/50 UVEX6568.1/51 UVEX6568.1/52 UVEX6568.2/35 UVEX6568.2/36 UVEX6568.2/37 UVEX6568.2/38 UVEX6568.2/39 UVEX6568.2/40 UVEX6568.2/41 UVEX6568.2/42 UVEX6568.2/43 UVEX6568.2/44 UVEX6568.2/45 UVEX6568.2/46 UVEX6568.2/47 UVEX6568.2/48 UVEX6568.2/49 UVEX6568.2/50 UVEX6568.2/51 UVEX6568.2/52 UVEX6568.3/35 UVEX6568.3/36 UVEX6568.3/37 UVEX6568.3/38 UVEX6568.3/39 UVEX6568.3/40 UVEX6568.3/41 UVEX6568.3/42 UVEX6568.3/43 UVEX6568.3/44 UVEX6568.3/45 UVEX6568.3/46 UVEX6568.3/47 UVEX6568.3/48 UVEX6568.3/49 UVEX6568.3/50 UVEX6568.3/51 UVEX6568.3/52
ESD safety shoes, Boa® Fit System, uvex 1 x-tended support, 6568
Chaussures et bottes
Sporty, multifunctional safety shoes are available in multiple widths.
  • Especially lightweight and flexible
  • Meets the ESD specification with volume resistance less than 35 megaohms
  • Breathable, perforated, high-tech microvelour upper material
  • Foamed polyurethane scuffcap helps increase product life of upper
  • Boa® fit system for an optimized fit, allowing precise and individual adjustment
The 100% metal-free uvex xenova® toe cap offers compact design, anatomical shape, good lateral stability, and no thermal conductivity properties. The metal-free, penetration-resistant insole is in accordance with the latest standards, and does not impact shoe flexibility. Medial and lateral heel support for the foot is provided through a specially designed side frame to protect against ankle injuries and impacts. Suitable for people allergic to chrome, as they are made from synthetic materials. All sole materials are free of silicones, plasticisers, and other paint wetting impairment substances.
Colour: Black/yellow
Certifications: EN 61340 (ESD); EN ISO 20345:2011 S1 P SRC certified.