Imprimer
OVS sampling tubes, ORBO™
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
20350 20029-U
SUPL20350EA 393 EUR
SUPL20350 20029-U
OVS sampling tubes, ORBO™
Échantillonneurs Échantillonneurs d'air
The OSHA Versatile Sampler (OVS) is a unique tube design with the adsorbent and filter combined in one tube for collection of both vapors and aerosols.
  • Packed with specially cleaned low background porous polymers
  • Most common sampling rate is 1 L/min to obtain a sample volume from 60 to 480 L
It′s most common use is for sampling pesticides but can also be used for other applications.