Imprimer

Poly(vinyl chloride), Selectophore™, poids moléculaire élevé ionophore, Supelco®

Marques : Merck
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5


M-Clarity™ (Niveau de qualité) = MQ100

81392-10G 81392-50G
81392-10GEA 62.3 EUR
81392-10G 81392-50G
Poly(vinyl chloride), Selectophore™, poids moléculaire élevé ionophore, Supelco®
Poly(vinyl chloride)

Poly(vinyl chloride), popular as PVC, is a polymer which is prepared from vinyl chloride monomer via polymerisation.


It can never be used as such and is mostly mixed with heat stabilizers, lubricants, plasticizers, fillers, and other additives. Due to its high chemical resistance property it finds wide application in construction and lining of chemical plants.


It has been used to fabricate Aluminum(III) selective potentiometric sensor from PVC matrix membrane which has morin ionophore

Formule: (CH₂CHCl)n
Numéro MDL: MFCD00084459
Numéro CAS: 9002-86-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=614408c19b2b88b8bdd5c8d87ed213e10edb36df&vid=5b2024d026198a37863b3a834560143159bcbaa1&prid=web&clid=SAPDAM

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR