Imprimer
(5-Dimethylamino)naphtalène-1-sulfonyle chlorure ≥99.0% (par HPLC), LiChropur™ réactif de dérivatisation, pour HPLC, Supelco®
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Danger
Synonyme(s): 5-(Dimethylamino)naphthalene-1-sulphonyl chloride
03641-100MG 03641-10X100MG
03641-100MGEA 42.3 EUR
03641-100MG 03641-10X100MG
(5-Dimethylamino)naphtalène-1-sulfonyle chlorure ≥99.0% (par HPLC), LiChropur™ réactif de dérivatisation, pour HPLC, Supelco®
(5-Dimethylamino)naphtalène-1-sulfonyle chlorure
Dansyl chloride is a fluorogenic reagent which is typically employed for the derivatization of amino acids.
Dansyl chloride has been used for the fluorogenic labeling of xenoestrogens, 4-nonylphenol and bisphenol A prior to their determination by high-performance liquid chromatography (HPLC) with fluorescence detection. It has also been used for the derivatization of icariin in human serum samples after oral administration of Epimedium decoction, followed by its determination using ultrasensitive LC-tandem mass spectrometry (LC-MS)and for the derivatization of antitumoral agent ES-285 in dog plasma followed by its analysis by HPLC equipped with ultraviolet and fluorescence detection.
Formule: C₁₂H₁₂ClNO₂S
Poids moléculaire: 269.75 g/mol
Température de stockage: Réfrigérateur
Numéro MDL: MFCD00003985
Numéro CAS: 605-65-2
UN: 3261
ADR: 8 II
Indice Merck: 13,02840