Imprimer
Supelpak™-2SV, Pore dim.: 90 Å (mean pore size) 20-60 mesh, Supelco®
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
M-Clarity™ (Niveau de Qualité) = MQ100
13673-U 13674-U 13682-U
13673-UEA 560 EUR
13673-U 13674-U 13682-U
Supelpak™-2SV, Pore dim.: 90 Å (mean pore size) 20-60 mesh, Supelco®
Supelpak™-2SV
Supelpak™-2SV is purified Amberlite® XAD2 that has been specially cleaned and tested for optimal performance in the capturing and extraction of semivolatile organics. Packaged in glass containers.
  • Matrix: Styrene-divinylbenzene
  • Pore size: ~0.65 mL/g pore volume
  • surface area: ~300 m2/g