Imprimer
Electrolyte filling solution for Ag/AgCl reference electrodes, Supelco®
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
60137-250ML 60137-1L
60137-250MLEA 27.4 EUR
60137-250ML 60137-1L
Electrolyte filling solution for Ag/AgCl reference electrodes, Supelco®
Electrolytes Electrode Filling Solutions
~3 M Potassium chloride solution saturated with silver chloride (~0,16 M AgCl).