Imprimer
Cyanine 680 succinimidyl ester colorant cyanine
Marques : Biotium
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
90048-1MG 90048-10MG
90048-1MGEA 308 EUR
90048-1MG 90048-10MG
Cyanine 680 succinimidyl ester colorant cyanine
Cyanine succinimidyl ester
Biotium offers high quality amine-reactive succinimidyl ester (SE) dyes used to label proteins, oligonucleotides, and nucleic acids.
  • Cyanine 680 is structurally identical to Cy®5.5 from GE Healthcare.
  • λEx/λEm (water) = 681/698 nm
  • Dark blue solid soluble in DMF and DMSO
  • Store at -20°C and protect from moisture and light
We offer Cyanine 555 (Cy®3), Cyanine 647 (Cy®5), Cyanine 680 (Cy®5.5), and Cyanine 750 (Cy®7) as high quality succinimidyl esters.
Also see our CF® Dye Succinimidyl Esters, available with more than 20 bright and photostable CF® Dyes.
Cy3, Cy5, Cy5.5, and Cy7 are registered trademarks of GE Healthcare.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6dd074a41b2d713e04861a949a0df4595d113a67&vid=a9d17b608f6d930c33c10c5820026a85a4b0ba7a&prid=web&clid=SAPDAM