Imprimer
Industrial gloves, uvex athletic allround
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6002806 6002807 6002808 6002809 6002810 6002811
UVEX6002806EA 45.8 EUR
UVEX6002806 UVEX6002807 UVEX6002808 UVEX6002809 UVEX6002810 UVEX6002811
Industrial gloves, uvex athletic allround
Gants
These lightweight knitted PA/elastan gloves with NBR foam coating on palm and fingertips are ideal for mechanical precision work.
  • Good mechanical abrasion resistance
  • Good grip in dry and slightly damp areas
  • Highly breathable
  • Very good tactile feel
  • Very good fit
  • Highly flexible
Areas of application: Precision assembly work, fine assembly work, inspection.
Certifications: EN 388:2016 (4122X)