Imprimer
Avantor® Hichrom, Connectors, PEEK Tees and Crosses
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HICHHI-034EA 83.5 EUR
HICHHI-034 HI-032 HICHHI-035
Avantor® Hichrom, Connectors, PEEK Tees and Crosses
Raccords et connecteurs
  • Use with one PEEK™ or stainless steel tubing
  • Maximum operating pressure 3,500psi (241 bar)
Informations de livraison: Includes three or four HI-505 fingertight fittings.